Hexaammineruthenium trichloride Ru ≥ 32,1% Ru ≥ 32.1% analytical reagent
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Hexaammineruthenium(III) chloride, Hexaammineruthenium (III) chloride
010511-01
010511-03
010511-06
AA10511-01EA
52
USD
AA10511-01
AA10511-03
AA10511-06
Hexaammineruthenium trichloride Ru ≥ 32,1% Ru ≥ 32.1% analytical reagent
Hexaammineruthenium trichloride
CAS No.: 14282-91-8
Molecular Formula: Ru(NH3)6Cl3
Formula Weight: 309.61
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Pale yellow
UN No.: 3288
MDL No.: MFCD00011478
Formula:
H₁₈Cl₃N₆Ru Storage Temperature: Refrigerator |
MDL Number:
MFCD00011478 CAS Number: 14282-91-8 EINECS: 238-176-8 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...