trans-Platin Pt ≥ 64,5%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: trans-Diamminedichloroplatinum (II), trans-Diammineplatinum (II) dichloride, trans-Dichlorodiammine platinum(II), trans-Dichlorodiamineplatinum(II)

010472-06 010472-01 010472-03
AA10472-06EA 944 USD
AA10472-06 AA10472-01 AA10472-03
trans-Platin Pt ≥ 64,5%
trans-Platin

MDL: MFCD00011623


CAS No.: 14913-33-8 Molecular Formula: Cl2H6N2Pt Formula Weight: 300.06 Physical Form: Crystalline Appearance: Yellow Odor: Odorless MDL No.: MFCD00011623

Formula: H₆Cl₂N₂Pt
Melting Pt: 340 °C (dec.)
Density: 3,746 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011623
CAS Number: 14913-33-8
EINECS: 238-980-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR