Zirconium ≥99.2% (metals basis excluding Hf), rods (max. 4.5% Hf), annealed, Diameter 12.7 mm (0.5 in)

Supplier: Thermo Scientific Chemicals

010442-BV 010442-BQ
AA10442-BVEA 320 USD
AA10442-BV AA10442-BQ
Zirconium ≥99.2% (metals basis excluding Hf), rods (max. 4.5% Hf), annealed, Diameter 12.7 mm (0.5 in)
Zirconium

MDL: MFCD00011303


Purity: 99.2+% CAS No.: 7440-67-7 Physical Form: ≈8.24g/cm, Temper: soft Odor: Odorless MDL No.: MFCD00011303

Formula: Zr
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR