Zinc ≥99.99% (metals basis), shot, Diameter 10mm (0.4in), Thickness 2 mm (0.08 in)

Supplier: Thermo Scientific Chemicals

010440-36 010440-22 010440-A3
AA10440-36EA 88.69 USD
AA10440-36 AA10440-22 AA10440-A3
Zinc ≥99.99% (metals basis), shot, Diameter 10mm (0.4in), Thickness 2 mm (0.08 in)
Zinc

MDL: MFCD00011291


Purity: 99.99% CAS No.: 7440-66-6 Odor: Odorless MDL No.: MFCD00011291

Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR