Zinc ≥99.98% (metals basis), foil, Width 30 cm (12 in), Thickness 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

010436-BW 010436-BS 010436-CB
AA10436-BWEA 95.62 USD
AA10436-BW AA10436-BS AA10436-CB
Zinc ≥99.98% (metals basis), foil, Width 30 cm (12 in), Thickness 0.25 mm (0.01 in)
Zinc

MDL: MFCD00011291


Purity: 99.98% CAS No.: 7440-66-6 Physical Form: ≈53g/30x10cm Odor: Odorless MDL No.: MFCD00011291

Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now

Specification Test Results

Appearance (Color) Gray
Form Foil
Comment 0.22 - 0.28 mm thick
Width 27 - 33 mm
Total Metal Impurities 0.02% max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR