Zinc ≥99.99% (metals basis), rods, Diameter 1.27 cm (0.5 in)

Supplier: Thermo Scientific Chemicals

010433-BV 010433-RM 010433-RS
AA10433-BVEA 99.7 USD
AA10433-BV AA10433-RM AA10433-RS
Zinc ≥99.99% (metals basis), rods, Diameter 1.27 cm (0.5 in)
Zinc

MDL: MFCD00011291


Purity: 99.99+% CAS No.: 7440-66-6 Physical Form: ≈9.03g/cm Odor: Odorless MDL No.: MFCD00011291

Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR