Ytterbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness 1.0 mm (0.04 in)

Supplier: Thermo Scientific Chemicals

010426-FF 010426-FI
AA10426-FFEA 178 USD
AA10426-FF AA10426-FI
Ytterbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness 1.0 mm (0.04 in)
Ytterbium

MDL: MFCD00011286 Notes: May contain up to 0.15% Calcium


Purity: 99.9% CAS No.: 7440-64-4 Physical Form: ≈4.4g/25x25mm, Packaged in mylar under argon Odor: Odorless MDL No.: MFCD00011286

Formula: Yb
Boiling Pt: 1427 °C (1013 hPa)
Melting Pt: 824 °C
Density: 6.977 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011286
CAS Number: 7440-64-4
EINECS: 231-173-2
Merck Index: 12,10239

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR