Tungsten ≥99.95% (metals basis), wire, Diameter 0.025 mm (0.001 in)

Supplier: Thermo Scientific Chemicals

010405-H4
AA10405-H4EA 220.15 USD
AA10405-H4 AA10405-H9
Tungsten ≥99.95% (metals basis), wire, Diameter 0.025 mm (0.001 in)
Tungsten

MDL: MFCD00011461


Purity: 99.95% CAS No.: 7440-33-7 Physical Form: ≈0.009g/m MDL No.: MFCD00011461

Formula: W
Boiling Pt: 5660 °C (1013 hPa)
Melting Pt: 3410 °C
Density: 19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR