Tungsten ≥99.9% (metals basis), Powder -100 mesh

Supplier: Thermo Scientific Chemicals
Warning

010402-36 010402-A3 010402-22
AA10402-36EA 185 USD
AA10402-36 AA10402-A3 AA10402-22
Tungsten ≥99.9% (metals basis), Powder -100 mesh
Tungsten

MDL: MFCD00011461


Purity: 99.9% CAS No.: 7440-33-7 Appearance: Grey UN No.: 3089 MDL No.: MFCD00011461

Formula: W
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
UN: 3089
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR