Tungsten ≥99.9% (metals basis), Powder APS 1-5 µm
Supplier: Thermo Scientific Chemicals
|
Warning
|
010400-09
010400-22
010400-36
010400-A3
AA10400-09EA
49.21
USD
AA10400-09
AA10400-22
AA10400-36
AA10400-A3
Tungsten ≥99.9% (metals basis), Powder APS 1-5 µm
Tungsten
MDL: MFCD00011461
Purity: 99.9%
CAS No.: 7440-33-7
Physical Form: Powder
Appearance: Grey
UN No.: 3089
MDL No.: MFCD00011461
Formula:
W Storage Temperature: Ambient |
MDL Number:
MFCD00011461 CAS Number: 7440-33-7 EINECS: 231-143-9 UN: 3089 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...