Thulium ≥99.9% (REO, rare earth oxide basis), foil, Thickness 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals
Danger

010375-FF 010375-FI
AA10375-FFEA 405 USD
AA10375-FF AA10375-FI
Thulium ≥99.9% (REO, rare earth oxide basis), foil, Thickness 0.25 mm (0.01 in)
Thulium

MDL: MFCD00011281


Purity: 99.9% CAS No.: 7440-30-4 Physical Form: ≈1.75g/25x25mm, Packaged under argon Appearance: Silver-grey Odor: Odorless UN No.: 3178 MDL No.: MFCD00011281

Formula: Tm
Storage Temperature: Ambient
MDL Number: MFCD00011281
CAS Number: 7440-30-4
EINECS: 231-140-2
UN: 3178
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR