Sodium 40% dispersion in oil , 99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
010340-56
010340-30
AA10340-56EA
372
USD
AA10340-56
AA10340-30
Sodium 40% dispersion in oil , 99% (metals basis)
Sodium
Purity: 99+%
CAS No.: 7440-23-5
Sensitivity: Air and Moisture Sensitive
Appearance: White to off-white
UN No.: 1391
MDL No.: MFCD00085307
Formula:
Na Storage Temperature: Ambient |
MDL Number:
MFCD00085307 CAS Number: 7440-23-5 EINECS: 231-132-9 UN: 1391 ADR: 4.3,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...