Platinum ≥99.95% (metals basis), wire, Diameter 0.025 mm (0.001 in)

Supplier: Thermo Scientific Chemicals

010292-G5 010292-H4 010292-G9
AA10292-G5EA 136.87 USD
AA10292-G5 AA10292-H4 AA10292-G9
Platinum ≥99.95% (metals basis), wire, Diameter 0.025 mm (0.001 in)
Platinum

MDL: MFCD00011179


Purity: 99.95% CAS No.: 7440-06-4 Physical Form: ≈0.01g/m Odor: Odorless MDL No.: MFCD00011179

Formula: Pt
Boiling Pt: 3827 °C (1013 hPa)
Melting Pt: 1769 °C
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR