Platinum, wire for ISA type R or S standard grade thermocouple, Diameter 0.0762 mm (0.003 in)

Supplier: Thermo Scientific Chemicals

010291-G2 010291-G6 010291-BW
AA10291-G2EA 91.3 USD
AA10291-G2 AA10291-G6 AA10291-BW
Platinum, wire for ISA type R or S standard grade thermocouple, Diameter 0.0762 mm (0.003 in)
Platinum

For ISA Type R or S Standard Grade Thermocouple


CAS No.: 7440-06-4 Physical Form: ≈0.05g/50cm Odor: Odorless MDL No.: MFCD00011179

Formula: Pt
Boiling Pt: 3827 °C (1013 hPa)
Melting Pt: 1769 °C
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR