Platinum, wire for ISA type R or S standard grade thermocouple, Diameter 0.2032 mm (0.008 in)

Supplier: Thermo Scientific Chemicals

010287-G1 010287-G5
AA10287-G1EA 372 USD
AA10287-G1 AA10287-G5
Platinum, wire for ISA type R or S standard grade thermocouple, Diameter 0.2032 mm (0.008 in)
Platinum

For ISA Type R or S Standard Grade Thermocouple


CAS No.: 7440-06-4 Physical Form: ≈0.69g/m Odor: Odorless MDL No.: MFCD00011179

Formula: Pt
Boiling Pt: 3827 °C (1013 hPa)
Melting Pt: 1769 °C
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR