Niobium ≥99.8% (metals basis excluding Ta), powder -60 mesh (max. 1% Ta)

Supplier: Thermo Scientific Chemicals

010276-36 010276-14 010276-22
AA10276-36EA 565 USD
AA10276-36 AA10276-14 AA10276-22
Niobium ≥99.8% (metals basis excluding Ta), powder -60 mesh (max. 1% Ta)
Niobium

MDL: MFCD00011126


Purity: 99.8% CAS No.: 7440-03-1 Odor: Odorless MDL No.: MFCD00011126

Formula: Nb
MW: 92.91 g/mol
Boiling Pt: 3700
Melting Pt: 2468
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5
UN: 3089
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR