Nickel ≥99.8% (metals basis), wire, Diameter 0.38 mm (0.015 in)

Supplier: Thermo Scientific Chemicals
Danger

010253-H4
AA10253-H4EA 148.18 USD
AA10253-H4 AA10253-H9
Nickel ≥99.8% (metals basis), wire, Diameter 0.38 mm (0.015 in)
Nickel

MDL: MFCD00011137


Purity: 99.8% CAS No.: 7440-02-0 Physical Form: ≈1.01g/m Odor: Odorless MDL No.: MFCD00011137

Formula: Ni
Boiling Pt: 3007 °C (1013 hPa)
Melting Pt: 1453 °C
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,III
Merck Index: 12,06582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR