Mercury ≥99.9998% (metals basis), Electronic Grade for the electronics industry

Supplier: Thermo Scientific Chemicals
Danger

010242-P2 010242-P9 010242-P5
AA10242-P2EA 344.17 USD
AA10242-P2 AA10242-P9 AA10242-P5
Mercury ≥99.9998% (metals basis), Electronic Grade for the electronics industry
Mercury

MDL: MFCD00011035


Purity: 99.9998% CAS No.: 7439-97-6 Appearance: Silver Odor: Odorless UN No.: 2809 MDL No.: MFCD00011035

Formula: Hg
Boiling Pt: 356.5 °C (1013 hPa)
Melting Pt: –39 °C
Density: 13.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011035
CAS Number: 7439-97-6
EINECS: 231-106-7
UN: 2809
ADR: 8,III
Merck Index: 13,05925

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR