Manganese ≥99.9% (metals basis), irregular pieces

Supplier: Thermo Scientific Chemicals

010236-30 010236-A1 010236-A7
AA10236-30EA 77.73 USD
AA10236-30 AA10236-A1 AA10236-A7
Manganese ≥99.9% (metals basis), irregular pieces
Manganese

MDL: MFCD00011111


Purity: 99.9% CAS No.: 7439-96-5 Physical Form: Broken plate Odor: Odorless MDL No.: MFCD00011111

Formula: Mn
Storage Temperature: Ambient
MDL Number: MFCD00011111
CAS Number: 7439-96-5
EINECS: 231-105-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR