Manganese ≥99.9% (metals basis), irregular pieces
Supplier: Thermo Scientific Chemicals
010236-30
010236-A1
010236-A7
AA10236-30EA
77.73
USD
AA10236-30
AA10236-A1
AA10236-A7
Manganese ≥99.9% (metals basis), irregular pieces
Manganese
MDL: MFCD00011111
Purity: 99.9%
CAS No.: 7439-96-5
Physical Form: Broken plate
Odor: Odorless
MDL No.: MFCD00011111
Formula:
Mn Storage Temperature: Ambient |
MDL Number:
MFCD00011111 CAS Number: 7439-96-5 EINECS: 231-105-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...