Magnesium ≥99.8% (metals basis), turnings
Supplier: Thermo Scientific Chemicals
|
Danger
|
010232-36
010232-22
AA10232-36EA
132.38
USD
AA10232-36
AA10232-22
Magnesium ≥99.8% (metals basis), turnings
Magnesium
MDL: MFCD00085308
Purity: 99.8%
CAS No.: 7439-95-4
Physical Form: 3.2mm (0.13in) wide
Appearance: Silver-grey
Odor: Odorless
UN No.: 1869
MDL No.: MFCD00085308
Formula:
Mg Storage Temperature: Ambient |
MDL Number:
MFCD00085308 CAS Number: 7439-95-4 EINECS: 231-104-6 UN: 1869 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...