Holmium ≥99.9% (REO, rare earth oxide basis), foil, Thickness 1.0 mm (0.04 in)

Supplier: Thermo Scientific Chemicals

010211-FF
AA10211-FFEA 256 USD
AA10211-FF AA10211-FI
Holmium ≥99.9% (REO, rare earth oxide basis), foil, Thickness 1.0 mm (0.04 in)
Holmium

MDL: MFCD00011049


Purity: 99.9% CAS No.: 7440-60-0 Physical Form: ≈5.5g/25x25mm, Packaged in mylar under argon Odor: Odorless MDL No.: MFCD00011049

Formula: Ho
Storage Temperature: Ambient
MDL Number: MFCD00011049
CAS Number: 7440-60-0
EINECS: 231-169-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR