Holmium ≥99.9% (REO, rare earth oxide basis), foil, Thickness 0.3 mm (0.01 in)
Supplier: Thermo Scientific Chemicals
010210-FF
010210-FI
AA10210-FFEA
287
USD
AA10210-FF
AA10210-FI
Holmium ≥99.9% (REO, rare earth oxide basis), foil, Thickness 0.3 mm (0.01 in)
Holmium
MDL: MFCD00011049
Purity: 99.9%
CAS No.: 7440-60-0
Physical Form: ≈1.65g/25x25mm, Packaged in mylar under argon
Odor: Odorless
MDL No.: MFCD00011049
Formula:
Ho Storage Temperature: Ambient |
MDL Number:
MFCD00011049 CAS Number: 7440-60-0 EINECS: 231-169-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...