Germanium ≥99.9999% (metals basis), ingot, zone refined 95% N type minimum

Supplier: Thermo Scientific Chemicals

010192-79 010192-TK
AA10192-79EA 1607 USD
AA10192-79 AA10192-TK
Germanium ≥99.9999% (metals basis), ingot, zone refined 95% N type minimum
Germanium

MDL: MFCD00085310


  • Electrical Resistivity: 40 Ohm-mc


Purity: 99.9999% CAS No.: 7440-56-4 Physical Form: Electrical Resistivity: 40 ohm-cm Odor: Odorless MDL No.: MFCD00085310

Formula: Ge
Boiling Pt: 2830 °C (1013 hPa)
Melting Pt: 937.2 °C
Density: 5.323 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085310
CAS Number: 7440-56-4
EINECS: 231-164-3
Merck Index: 12,04415

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR