Gallium ≥99.99% (metals basis), ingot

Supplier: Thermo Scientific Chemicals
Warning

010188-18 010188-30 010188-36 010188-14
AA10188-18EA 352 USD
AA10188-18 AA10188-30 AA10188-36 AA10188-14
Gallium ≥99.99% (metals basis), ingot
Formula: Ga
Boiling Pt: 2403 °C (1013 hPa)
Melting Pt: 29 °C
Density: 5.904 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00134045
CAS Number: 7440-55-3
EINECS: 231-163-8
UN: 2803
ADR: 8,III
Merck Index: 12,04364

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR