Gallium ≥99.99% (metals basis), ingot
Supplier: Thermo Scientific Chemicals
|
Warning
|
010188-18
010188-30
010188-36
010188-14
AA10188-18EA
352
USD
AA10188-18
AA10188-30
AA10188-36
AA10188-14
Gallium ≥99.99% (metals basis), ingot
Gallium
MDL: MFCD00134045
Purity: 99.99%
CAS No.: 7440-55-3
Molecular Formula: Requires overnight shipping with cold pack
Storage Temperature: 2°C to 8°C
Appearance: Silver grey
MDL No.: MFCD00134045
Formula:
Ga Boiling Pt: 2403 °C (1013 hPa) Melting Pt: 29 °C Density: 5.904 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00134045 CAS Number: 7440-55-3 EINECS: 231-163-8 UN: 2803 ADR: 8,III Merck Index: 12,04364 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...