Gallium ≥99.99999% (metals basis), ingot
Supplier: Thermo Scientific Chemicals
010186-36
010186-30
010186-18
010186-14
AA10186-36EA
3122
USD
AA10186-36
AA10186-30
AA10186-18
AA10186-14
Gallium ≥99.99999% (metals basis), ingot
Gallium
MDL: MFCD00134045
Purity: 99.99999%
CAS No.: 7440-55-3
Molecular Formula: Requires overnight shipping with cold pack
Storage Temperature: 2°C to 8°C
Appearance: Silver
MDL No.: MFCD00134045
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...