Gallium ≥99.9999% (metals basis), ingot
Supplier: Thermo Scientific Chemicals
010185-18
AA10185-18EA
406
USD
AA10185-18
Gallium ≥99.9999% (metals basis), ingot
MDL: MFCD00134045
Purity: 99.9999%
CAS No.: 7440-55-3
Storage Temperature: 2°C to 8°C
Appearance: Silver grey
UN No.: 2803
MDL No.: MFCD00134045
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...