Chromium ≥99% (metals basis), powder -100 mesh

Supplier: Thermo Scientific Chemicals
Warning

010147-A3 010147-22 010147-36 010147-09
AA10147-A3EA 796 USD
AA10147-A3 AA10147-22 AA10147-36 AA10147-09
Chromium ≥99% (metals basis), powder -100 mesh
Chromium

MDL: MFCD00010944


Purity: 99% CAS No.: 7440-47-3 Physical Form: Apparent density 2.0-3.0g/cm3 Appearance: Silver Odor: Odorless UN No.: 3089 MDL No.: MFCD00010944

Formula: Cr
Storage Temperature: Ambient
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
UN: 3089
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR