Bismuth ≥99.5% (metals basis), powder -325 mesh
Supplier: Thermo Scientific Chemicals
|
Warning
|
010111-22
010111-36
AA10111-22EA
65.68
USD
AA10111-22
AA10111-36
Bismuth ≥99.5% (metals basis), powder -325 mesh
Bismuth
MDL: MFCD00134033
Purity: 99.5%
CAS No.: 7440-69-9
Physical Form: Apparent density 3.2-4.0g/cm3
Odor: Odorless
UN No.: 3089
MDL No.: MFCD00134033
Formula:
Bi Density: 9.78 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134033 CAS Number: 7440-69-9 EINECS: 231-177-4 UN: 3089 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...