Bismuth ≥99.998% (metals basis), needles, Length 1-5 cm (0.4-2 in)

Supplier: Thermo Scientific Chemicals

010109-22 010109-36 010109-A3
AA10109-22EA 71.3 USD
AA10109-22 AA10109-36 AA10109-A3
Bismuth ≥99.998% (metals basis), needles, Length 1-5 cm (0.4-2 in)
Bismuth

MDL: MFCD00134033


Purity: 99.998% CAS No.: 7440-69-9 Physical Form: Approximately 1.5mm diameter Odor: Odorless MDL No.: MFCD00134033

Formula: Bi
Boiling Pt: 1490 °C (1013 hPa)
Melting Pt: 271…272 °C
Density: 9.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
Merck Index: 12,01297

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR