Beryllium ≥99% (metals basis), powder -325 mesh

Supplier: Thermo Scientific Chemicals
Danger

010108-22 010108-14
AA10108-22EA 4000 USD
AA10108-22 AA10108-14
Beryllium ≥99% (metals basis), powder -325 mesh
Beryllium

MDL: MFCD00134032


Purity: 99+% CAS No.: 7440-41-7 Odor: Odorless UN No.: 1567 MDL No.: MFCD00134032

Formula: Be
Storage Temperature: Ambient
MDL Number: MFCD00134032
CAS Number: 7440-41-7
EINECS: 231-150-7
UN: 1567
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR