Aluminium ≥99.5% (metals basis), lumps

Supplier: Thermo Scientific Chemicals

010092-30 010092-A7 010092-A1
AA10092-30EA 53.2 USD
AA10092-30 AA10092-A7 AA10092-A1
Aluminium ≥99.5% (metals basis), lumps
Aluminium

MDL: MFCD00134029


Purity: 99.5% CAS No.: 7429-90-5 Physical Form: ≈17g/lump Odor: Odorless MDL No.: MFCD00134029

Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR