Bis(acetonitrile)palladium(II) dichloride (40,5% Pd)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Bis(acetonitrile)-palladium(II) chloride, Palladium(II) chloride diacetonitrile complex, Dichlorobis(acetonitrile)palladium(II)
For olefin isomerization, preparation of acetals and hemiacetal esters.
Soluble in acetonitrile, acetone, chloroform.
CAS No.: 14592-56-4
Molecular Formula: PdCl2(CH3CN)2
Formula Weight: 259.41
Physical Form: Powder
Odor: Odorless
Solubility: Soluble in acetonitrile, acetone, chloroform
UN No.: 3439
MDL No.: MFCD00013122
Applications: Olefin isomerization, preparation of acetals and hemiacetal esters
Formula:
C₄H₆Cl₂N₂Pd Storage Temperature: Ambient |
MDL Number:
MFCD00013122 CAS Number: 14592-56-4 EINECS: 238-637-3 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...