Samarium ≥99.9% (REO, rare earth oxide basis), foil, Thickness 0.62 mm (0.024 in)

Supplier: Thermo Scientific Chemicals

000610-FF 000610-FI
AA00610-FFEA 323 USD
AA00610-FF AA00610-FI
Samarium ≥99.9% (REO, rare earth oxide basis), foil, Thickness 0.62 mm (0.024 in)
Samarium

MDL: MFCD00011233


Purity: 99.9% CAS No.: 7440-19-9 Physical Form: ≈2.9g/25x25mm, Packaged in mylar under argon Odor: Odorless MDL No.: MFCD00011233

Formula: Sm
MW: 150.36 g/mol
Boiling Pt: 1900
Melting Pt: 1076.85
Density: 7.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011233
CAS Number: 7440-19-9
EINECS: 231-128-7
Merck Index: 12,08498

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR