Tungsten ≥99.9% (metals basis), slug, Length 6.4 mm (0.25 in), Diameter 17.5 mm (0.689 in)

Supplier: Thermo Scientific Chemicals

000525-KG 000525-KI
AA00525-KGEA 131 USD
AA00525-KG AA00525-KI
Tungsten ≥99.9% (metals basis), slug, Length 6.4 mm (0.25 in), Diameter 17.5 mm (0.689 in)
Tungsten

MDL: MFCD00011461


Purity: 99.9% CAS No.: 7440-33-7 Physical Form: ≈30g/slug MDL No.: MFCD00011461

Formula: W
Boiling Pt: 5660 °C (1013 hPa)
Melting Pt: 3410 °C
Density: 19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR