Zirconium ≥99.95% (metals basis excluding Hf), wire (max. 0.25% Hf), Diameter 0.127 mm (0.005 in)
Supplier: Thermo Scientific Chemicals
000419-G1
000419-G5
AA00419-G1EA
73.2
USD
AA00419-G1
AA00419-G5
Zirconium ≥99.95% (metals basis excluding Hf), wire (max. 0.25% Hf), Diameter 0.127 mm (0.005 in)
Zirconium
MDL: MFCD00011303
Purity: 99.95%
CAS No.: 7440-67-7
Physical Form: ≈0.082g/m
Odor: Odorless
MDL No.: MFCD00011303
Formula:
Zr Storage Temperature: Ambient |
MDL Number:
MFCD00011303 CAS Number: 7440-67-7 EINECS: 231-176-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...