Dysprosium ≥99.8% (REO, rare earth oxide basis), ingot

Supplier: Thermo Scientific Chemicals

000102-22 000102-14
AA00102-22EA 344 USD
AA00102-22 AA00102-14
Dysprosium ≥99.8% (REO, rare earth oxide basis), ingot
Dysprosium

MDL: MFCD00010982


Purity: 99.8% CAS No.: 7429-91-6 Physical Form: Packaged in air stable poly bottle Odor: Odorless MDL No.: MFCD00010982

Formula: Dy
Storage Temperature: Ambient
MDL Number: MFCD00010982
CAS Number: 7429-91-6
EINECS: 231-073-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR