Boc-β-cyano-D-Ala-OH

Supplier: BACHEM AMERICAS INC

4004994.0001
A-1540.0001BAEA 196.08 USD
A-1540.0001BA A-1540.0005BA
Boc-β-cyano-D-Ala-OH
Boc-beta-cyano-D-Ala-OH

Sequence: Boc-β-cyano-D-Ala-OH


Besides Ala derivatives, Bachem offers a number of derivatives of β-substituted alanines, amongst them: t-Butylalanine Cyanoalanine Cyclohexylalanine (Cha) Cyclopropylalanine 2-Quinolylalanine β-Halogenated Alanines
β-Nitrile of aspartic acid, formed by dehydration of the Asn carboxamide moiety.

Formula: C₉H₁₄N₂O₄
Storage Temperature: Freezer
MDL Number: MFCD00236830
CAS Number: 184685-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR