Nɑ-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine
Supplier: BACHEM AMERICAS INC
Synonyms:
Boc-Orn(Z)-OH, Nɑ-Boc-Nδ-Cbz-L-ornithine, Nδ-Benzyloxycarbonyl-Nα-Boc-L-ornithine
4005026.0025
4005026.0100
A-1490.0025BAEA
167.37
USD
A-1490.0025BA
A-1490.0100BA
Nɑ-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine
Nalpha-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine
Sequence: Boc-Orn(Z)-OH
Formula:
C₁₈H₂₆N₂O₆ Storage Temperature: Ambient |
MDL Number:
MFCD00038259 CAS Number: 2480-93-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...