Anti-CD3 Rabbit Monoclonal Antibody [clone: SP7]

Supplier: Diagnostic Biosystems

RMAB005
99991-136EA 1164.49 USD
99991-136
Anti-CD3 Rabbit Monoclonal Antibody [clone: SP7]
Antibodies
Type:
Antigen: CD3
Clonality: Monoclonal
Clone: SP7
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR