Taq Extender PCR Additive, Agilent Technologies
Supplier: AGILENT TECH
600148
99902-292EA 261.08 USD
99902-292
Taq Extender PCR Additive, Agilent Technologies
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
Improves reliability and yield of conventional Taq-based PCR amplifications.
  • Improves length, yield, and reliability of PCR; Enhances efficiency of template extension by Taq DNA polymerase
  • Improves amplification of difficult targets
Taq Extender PCR additive improves the reliability and yield of conventional Taq-based PCR amplifications. Taq Extender additive increases the efficiency of Taq DNA polymerase extension reactions during each cycle of PCR by reducing mismatch pausing, resulting in a greater percentage of completed extension reactions.
Caution: For research use only. Not for use in diagnostic procedures.