IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99% (by TLC)

Supplier: AGILENT TECH
Warning

300127
99901-500EA 99.88 USD
99901-500
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99% (by TLC)
IPTG (Isopropyl-β-D-thiogalactopyranoside)

IPTG is for use in combination with X-gal to detect β-galactosidase activity in blue/white selection assays.


Caution: For Research Use Only. Not for Use in Diagnostic Procedures.

Formula: C₉H₁₈O₅S
Melting Pt: 112...114 °C
Storage Temperature: Freezer
MDL Number: MFCD00063273
CAS Number: 367-93-1

Order Now

Specification Test Results

Purity ≥99% (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR