Taq2000 DNA Polymerase, Agilent Technologies
Supplier: AGILENT TECH
200435
97066-442EA 805.07 USD
97066-442 99900-168 99900-532 ST600195
Taq2000 DNA Polymerase, Agilent Technologies
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Highly purified, recombinant Taq DNA Polymerase cloned from thermophilic eubacteria, Thermus aquaticus.
  • Ultrapure cloned Taq DNA polymerase
  • Virtually eliminates background artifacts
  • Minimizes smearing in long PCR
  • Most thermostable Taq DNA polymerase
Taq2000 DNA Polymerase is a highly purified, recombinant Taq DNA polymerase cloned from the thermophilic eubacteria, Thermus aquaticus.
Caution: For research use only. Not for use in diagnostic procedures.