Super Erecta Shelf® Solid Shelving, All-Stainless Steel, Metro™
Supplier: Metro
1436NFS 1824NFS 2130NFS 2142NFS 2442NFS 1424NFS 1430NFS 1442NFS 1460NFS 1848NFS 2124NFS 2136NFS 2160NFS 2424NFS 2448NFS 1830NFS 1842NFS 2460NFS 1860NFS 2148NFS 2430NFS 2436NFS 1448NFS 1836NFS
98000-684EA 487.91 USD
98000-684 98000-692 98000-706 98000-710 98000-722 98000-680 98000-682 98000-686 98000-690 98000-700 98000-704 98000-708 98000-714 98000-716 98000-724 98000-694 98000-698 98000-726 98000-702 98000-712 98000-718 98000-720 98000-688 98000-696
Super Erecta Shelf® Solid Shelving, All-Stainless Steel, Metro™
Furniture Shelves
Solid shelving is available in four widths and six lengths, all stainless solid shelf with stainless corners.
  • All-stainless, 18-gauge steel construction with ¹/₈" (3 mm) ship's edge detail helps contain spills
  • All-stainless design, including stainless cast corners, can be used in cart wash and autoclave environments
Shelves up to 121.9 cm (48") in length can support up to 363 kg (800 lbs.) per level. Shelves longer than 121.9 cm (48") can support up to 272 kg (600 lbs.) per level.
Note: The 35.6 cm (14") wide shelves require the use of footplates (see 19766-814 series) when used as a freestanding unit. The actual length of the shelves is 3.2 mm (¹/₈") shorter than the nominal dimension shown. Stationary posts over 189.6 cm high and mobile posts over 138.6 cm high are not for use with 35.6 cm wide shelves. The height of the chrome-plated and stainless steel stationary posts includes a leveling bolt and cap.
Shelves meet MIL-S-40144E specifications.
Type 304 18-gauge stainless steel shelves provide maximum protection in corrosive environments. Solid shelves have a 3 mm (¹/₈") ship's edge detail and stainless cast corners. Easy shelf adjustment is possible through the use of SiteSelect® stationary or mobile posts, which allow shelves to be readjusted and snapped into place at precise 2.5 cm (1") increments along the entire height of the post. An easily identifiable double groove is positioned every 20.3 cm (8") to further aid shelf alignment. Stationary posts are fitted with adjustable stainless steel leveling bolts for use on uneven surfaces.
Ordering information: Mobile posts require the purchase of stem casters. Stainless steel posts (see 19766-068 series), stem casters (see 19766-236 series), and shelf dividers and ledges (see 22233-212 series) are available separately. Each shelf includes four pairs of plastic split sleeves for securing shelf to posts.