Cleanroom Storage Rack, Stainless Steel, Bandy
Supplier: Bandy
DISP60001
98000-584EA 3346.19 USD
98000-584
Cleanroom Storage Rack, Stainless Steel, Bandy
Furniture Gowning Racks
Twelve-compartment rack is ideal for storage of disposable cleanroom items such as gloves, booties, or bouffant caps.
  • All surface perforated
  • Perforations allow appropriate air flow
  • Adjustable leveling feet
Constructed of 16-gauge, Type 304 stainless steel with a No. 4 finish.