VWR® Electronic Stem Thermometer, Calibrated, Blunt Tip, Stainless Steel, 120 mm Probe
Supplier: VWR International
60900-2111
98000-198EA 47.73 USD
98000-198
VWR® Electronic Stem Thermometer, Calibrated, Blunt Tip, Stainless Steel, 120 mm Probe
Thermometers Digital Thermometers
Digital stem thermometer is suitable for field studies, wet lab applications, wash down areas, food testing and tanks.
  • Will read temperatures in liquids, semi-solids and gases
  • Stainless-steel probe easily reaches into cuvettes, test tubes, flasks, beakers and more
  • Includes accredited calibration documentation that meets the requirements of ISO 17025
  • Individually serialized, CE marked
Certifications: Tested and calibrated in exclusive triple accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.
Delivery information: Includes multi-language instructions (English, French, German, Italian, Portuguese, and Spanish).
Caution: These electronic stem thermometers are not suitable for solids such as frozen items.