VWR® Electronic Stem Thermometer, Calibrated, Stainless Steel, 127 mm Probe
Supplier: VWR International
98000-156
98000-156EA 65.7 USD
98000-156
VWR® Electronic Stem Thermometer, Calibrated, Stainless Steel, 127 mm Probe
Thermometers Digital Thermometers
Digital stem thermometer is suitable for field studies, wet lab applications, wash down areas, food testing and tanks.
  • Will read temperatures in liquids, semi-solids and gases
  • Stainless-steel probe easily reaches into cuvettes, test tubes, flasks, beakers and more
  • Includes accredited calibration documentation that meets the requirements of ISO 17025
  • Individually serialized, CE marked
Certifications: Tested and calibrated in exclusive triple accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.
Delivery information: Includes multi-language instructions (English, French, German, Italian, Portuguese, and Spanish).
Caution: These electronic stem thermometers are not suitable for solids such as frozen items.