Buchi® V-850 and V-855 Vacuum Controllers
Supplier: Buchi
044989
98000-024EA 95.68 USD
98000-024
Buchi® V-850 and V-855 Vacuum Controllers
Pumps Vacuum Controllers
Vacuum controllers can be universally used for the display, adjustment, and control of vacuum operation in conjunction with V-700 and V-710 vacuum pumps, as well as Rotavapor® R-210 and R-215 evaporators
The easy-to-operate units are equipped with a single control knob and a large graphic display screen with intuitive menu guidance and six selectable language options. Both controllers feature a solvent database, a timer function for process termination, and a LabVac function for control of laboratory vacuum systems. Units are equipped with a USB port and RS-232/RS-485 interfaces.
The V-855 controller is distinguished by additional features such as a gradient and repetition function for gentle distillation under strict requirements, gentle autodistillation for varying solvents and mixtures, and the EasyVac function, which provides automatic evaporation based on vapor pressure detection.
These compact and chemical-resistant units are suitable for a wide array of laboratory applications.