Linearity Blood Gas, AUDIT Microcontrols
Supplier: AUDIT MICROCONTROLS
K727M-5
97068-912PK 603.68 USD
97068-912
Linearity Blood Gas, AUDIT Microcontrols
Controls and Calibration Verification Materials
Blood gas calibration verification is assayed quality control material consisting of five levels of buffers and salts in a bovine based material and equilibrated with known levels of carbon dioxide (CO₂), oxygen (O₂), and nitrogen.
A quality control material intended for use in the quantitative verification of calibration and reportable range.